Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02346185_g1
          See other RT1-CE1 GE Assays ›
          Gene Symbol:
          RT1-CE1
          Gene Name:
          RT1 class I, locus1
          Gene Aliases:
          RT1-C/E1
          Chromosome Location:
          Chr.20: 4638235 - 4792390 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02346185_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RT1-CE1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008832.2 NP_001008832.1 5 - 6 1046 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001819.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P513029 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700287220 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701092543 - -
          Agilent Rat cDNA Microarray (G4105A) 700287220 - -
          Agilent Rat cDNA Microarray (G4105A) 701092543 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10831111 - -
          Affymetrix Rat Gene 1.0 ST Array 10833947 - -
          Affymetrix Rat Gene 1.0 ST Array 10833946 - -
          Affymetrix Rat Gene 1.0 ST Array 10831109 - -
          Affymetrix Rat Gene 1.0 ST Array 10831108 - -
          Affymetrix Rat Gene 1.0 ST Array 10833948 - -
          Affymetrix Rat Gene 1.0 ST Array 10831113 - -
          Affymetrix Rat Gene 1.0 ST Array 10833949 - -
          Affymetrix Rat Gene 1.0 ST Array 10831112 - -
          Affymetrix Rat Gene 1.0 ST Array 10831110 - -
          Affymetrix Rat Gene 1.0 ST Array 10833945 - -
          Affymetrix Rat Gene 1.0 ST Array 10833951 - -
          Affymetrix Rat Gene 1.1 ST Array 10831105 - -
          Affymetrix Rat Gene 1.1 ST Array 10833944 - -
          Rat Genome 230 2.0 Array 1371267_PM_at - -
          Rat Genome 230 2.0 Array 1371267_at - -
          Affymetrix Rat Clariom S Assay PSR2000014981.rn.1 TC2000001819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014980.rn.1 TC2000001819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014983.rn.1 TC2000001819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014968.rn.1 TC2000001819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014982.rn.1 TC2000001819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014979.rn.1 TC2000001819.rn.1 RTA TC
          Rat Expression Set 230 1371267_PM_at - -
          Rat Expression Set 230 1371267_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833944 - -
          Affymetrix Rat Gene 1.0 ST Array 10831105 - -
          Rat Genome U34 Set M64795_i_at - -
          Rat Genome U34 Set M64795_f_at - -
          Rat Genome U34 Set rc_AA899862_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21409488 - -
          ABI Rat Genome Survey Microarray 21534368 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376633 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline