Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02346575_m1
          See other NT5DC2 GE Assays ›
          Gene Symbol:
          Nt5dc2
          Gene Name:
          5'-nucleotidase domain containing 2
          Gene Aliases:
          RGD1305524
          Chromosome Location:
          Chr.16: 7212488 - 7220541 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02346575_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nt5dc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009271.1 NP_001009271.1 10 - 11 1166 79
          XM_008770986.1 XP_008769208.1 10 - 11 3208 79
          GenBank mRNA AY569011.1 - 10 - 11 1159 79
          BC088431.1 - 10 - 11 1166 7124640 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000093.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nt5dc2

          Panther Classification:

          Molecular Function -

          nucleotide phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          dephosphorylation
          5'-nucleotidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005043 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005043 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005043 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11350 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1002452 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786636 - -
          Affymetrix Rat Gene 1.0 ST Array 10786635 - -
          Affymetrix Rat Gene 1.0 ST Array 10786632 - -
          Affymetrix Rat Gene 1.0 ST Array 10786627 - -
          Affymetrix Rat Gene 1.0 ST Array 10786639 - -
          Affymetrix Rat Gene 1.0 ST Array 10786629 - -
          Affymetrix Rat Gene 1.0 ST Array 10786631 - -
          Affymetrix Rat Gene 1.0 ST Array 10786634 - -
          Affymetrix Rat Gene 1.0 ST Array 10786628 - -
          Affymetrix Rat Gene 1.0 ST Array 10786633 - -
          Affymetrix Rat Gene 1.0 ST Array 10786630 - -
          Affymetrix Rat Gene 1.0 ST Array 10786638 - -
          Affymetrix Rat Gene 1.0 ST Array 10786637 - -
          Affymetrix Rat Gene 1.0 ST Array 10786626 - -
          Affymetrix Rat Gene 1.1 ST Array 10786624 - -
          Rat Genome 230 2.0 Array 1371849_at - -
          Rat Genome 230 2.0 Array 1371849_PM_at - -
          Affymetrix HT Rat Focus Array 1371849_at - -
          Affymetrix HT Rat Focus Array 1371849_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600000982.rn.1 TC1600000093.rn.1 RTA TC
          Rat Expression Set 230 1371849_PM_at - -
          Rat Expression Set 230 1371849_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786624 - -
          Rat Genome U34 Set rc_AI111551_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21195405 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356169 - -
          RatRef-12 Expression BeadChip ILMN_57348 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline