Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02346659_m1
          See other MRPL38 GE Assays ›
          Gene Symbol:
          Mrpl38
          Gene Name:
          mitochondrial ribosomal protein L38
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 104682372 - 104691357 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02346659_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009369.2 NP_001009369.2 6 - 7 727 133
          XM_008768372.1 XP_008766594.1 6 - 7 1515 133
          GenBank mRNA BC087096.1 - 6 - 7 724 7191805 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004142.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl38

          Panther Classification:

          Molecular Function -

          protease inhibitor

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159896 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047401 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047401 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047401 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P483313 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10749133 - -
          Affymetrix Rat Gene 1.0 ST Array 10749135 - -
          Affymetrix Rat Gene 1.0 ST Array 10749138 - -
          Affymetrix Rat Gene 1.0 ST Array 10749136 - -
          Affymetrix Rat Gene 1.0 ST Array 10749137 - -
          Affymetrix Rat Gene 1.0 ST Array 10749139 - -
          Affymetrix Rat Gene 1.0 ST Array 10749140 - -
          Affymetrix Rat Gene 1.0 ST Array 10749134 - -
          Affymetrix Rat Gene 1.0 ST Array 10749141 - -
          Affymetrix Rat Gene 1.1 ST Array 10749132 - -
          Rat Genome 230 2.0 Array 1383618_at - -
          Rat Genome 230 2.0 Array 1383618_PM_at - -
          Affymetrix HT Rat Focus Array 1383618_PM_at - -
          Affymetrix HT Rat Focus Array 1383618_at - -
          Affymetrix Rat Clariom S Assay PSR1000037635.rn.1 TC1000004142.rn.1 RTA TC
          Rat Expression Set 230 1383618_at - -
          Rat Expression Set 230 1383618_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10749132 - -
          Rat Genome U34 Set rc_AA891689_g_at - -
          Rat Genome U34 Set rc_AA891689_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21351090 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline