Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009506.1 | NP_001009506.1 | 1 | 856 | 115 | |
GenBank mRNA | AY510344.1 | - | 1 | 856 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |