Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02346978_m1
          See other B4GALT3 GE Assays ›
          Gene Symbol:
          B4galt3
          Gene Name:
          UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 89643852 - 89649880 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02346978_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B4galt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009539.1 NP_001009539.1 6 - 7 1079 170
          XM_006250253.2 XP_006250315.1 6 - 7 1110 170
          XM_006250254.2 XP_006250316.1 6 - 7 1230 170
          GenBank mRNA BC061812.1 - 6 - 7 1079 5598658 170

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000755.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B4galt3

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase activity
          N-acetyllactosamine synthase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P487031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818050 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P487031 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P487031 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P487031 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P487031 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3694 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765549 - -
          Affymetrix Rat Gene 1.0 ST Array 10765544 - -
          Affymetrix Rat Gene 1.0 ST Array 10765548 - -
          Affymetrix Rat Gene 1.0 ST Array 10765547 - -
          Affymetrix Rat Gene 1.0 ST Array 10765543 - -
          Affymetrix Rat Gene 1.0 ST Array 10765545 - -
          Affymetrix Rat Gene 1.0 ST Array 10765546 - -
          Affymetrix Rat Gene 1.0 ST Array 10765550 - -
          Affymetrix Rat Gene 1.1 ST Array 10765542 - -
          Rat Genome 230 2.0 Array 1367487_at - -
          Rat Genome 230 2.0 Array 1367487_PM_at - -
          Affymetrix HT Rat Focus Array 1367487_at - -
          Affymetrix HT Rat Focus Array 1367487_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300005244.rn.1 TC1300000755.rn.1 RTA TC
          Rat Expression Set 230 1367487_at - -
          Rat Expression Set 230 1367487_PM_at - -
          Rat Expression Set 230 1367487_at - -
          Rat Expression Set 230 1367487_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765542 - -
          Rat Genome U34 Set rc_AI012277_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21931743 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367011 - -
          RatRef-12 Expression BeadChip ILMN_53804 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline