Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347236_m1
          See other KIF22 GE Assays ›
          Gene Symbol:
          Kif22
          Gene Name:
          kinesin family member 22
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 198461406 - 198476430 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02347236_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kif22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009645.1 NP_001009645.1 11 - 12 1678 110
          GenBank mRNA BC088421.1 - 11 - 12 1678 7123493 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006687.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kif22

          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          microtubule-based movement
          sister chromatid cohesion
          mitotic metaphase plate congression
          metabolic process
          DNA binding
          microtubule motor activity
          ATP binding
          microtubule binding
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P108200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50478 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108200 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108200 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P108200 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P227065 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41227 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8379 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63790 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725932 - -
          Affymetrix Rat Gene 1.0 ST Array 10725928 - -
          Affymetrix Rat Gene 1.0 ST Array 10725934 - -
          Affymetrix Rat Gene 1.0 ST Array 10725940 - -
          Affymetrix Rat Gene 1.0 ST Array 10725937 - -
          Affymetrix Rat Gene 1.0 ST Array 10725938 - -
          Affymetrix Rat Gene 1.0 ST Array 10725933 - -
          Affymetrix Rat Gene 1.0 ST Array 10725935 - -
          Affymetrix Rat Gene 1.0 ST Array 10725931 - -
          Affymetrix Rat Gene 1.0 ST Array 10725930 - -
          Affymetrix Rat Gene 1.0 ST Array 10725939 - -
          Affymetrix Rat Gene 1.0 ST Array 10725929 - -
          Affymetrix Rat Gene 1.0 ST Array 10725941 - -
          Affymetrix Rat Gene 1.0 ST Array 10725936 - -
          Affymetrix Rat Gene 1.1 ST Array 10725927 - -
          Rat Genome 230 2.0 Array 1372516_at - -
          Rat Genome 230 2.0 Array 1372516_PM_at - -
          Affymetrix HT Rat Focus Array 1372516_at - -
          Affymetrix HT Rat Focus Array 1372516_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100052698.rn.1 TC0100006687.rn.1 RTA TC
          Rat Expression Set 230 1372516_PM_at - -
          Rat Expression Set 230 1372516_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725927 - -
          Rat Genome U34 Set rc_AA965280_at - -
          Rat Genome U34 Set rc_AA851392_at - -
          Rat Genome U34 Set rc_AA851392_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21241288 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358183 - -
          RatRef-12 Expression BeadChip ILMN_63168 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline