Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347432_m1
          See other FASTKD2 GE Assays ›
          Gene Symbol:
          Fastkd2
          Gene Name:
          FAST kinase domains 2
          Gene Aliases:
          RGD1307883
          Chromosome Location:
          Chr.9: 70448386 - 70471420 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02347432_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fastkd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009673.1 NP_001009673.1 9 - 10 1918 146
          XM_006245050.1 XP_006245112.1 10 - 11 2075 146
          GenBank mRNA BC088416.1 - 9 - 10 1918 7112913 146
          FQ218822.1 - 5 - 6 723 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000796.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fastkd2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          cellular respiration
          protein kinase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P667782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116930 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P667782 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P667782 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P667782 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P667782 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89323 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87062 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923914 - -
          Affymetrix Rat Gene 1.0 ST Array 10923911 - -
          Affymetrix Rat Gene 1.0 ST Array 10923910 - -
          Affymetrix Rat Gene 1.0 ST Array 10923913 - -
          Affymetrix Rat Gene 1.0 ST Array 10923912 - -
          Affymetrix Rat Gene 1.0 ST Array 10923907 - -
          Affymetrix Rat Gene 1.0 ST Array 10923905 - -
          Affymetrix Rat Gene 1.0 ST Array 10923906 - -
          Affymetrix Rat Gene 1.0 ST Array 10923916 - -
          Affymetrix Rat Gene 1.0 ST Array 10923909 - -
          Affymetrix Rat Gene 1.0 ST Array 10923915 - -
          Affymetrix Rat Gene 1.1 ST Array 10923904 - -
          Rat Genome 230 2.0 Array 1379337_at - -
          Rat Genome 230 2.0 Array 1379337_PM_at - -
          Affymetrix HT Rat Focus Array 1379337_at - -
          Affymetrix HT Rat Focus Array 1379337_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900006481.rn.1 TC0900000796.rn.1 RTA TC
          Rat Expression Set 230 1379337_PM_at - -
          Rat Expression Set 230 1379337_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923904 - -
          Rat Genome U34 Set rc_AI232281_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21828820 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358502 - -
          RatRef-12 Expression BeadChip ILMN_50404 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline