Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347909_mH
          See other HNRNPM GE Assays ›
          Gene Symbol:
          Hnrnpm
          Gene Name:
          heterogeneous nuclear ribonucleoprotein M
          Gene Aliases:
          Hnrpm, Hnrpm4
          Chromosome Location:
          Chr.7: 18516253 - 18555438 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02347909_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrnpm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109911.1 NP_001103381.1 12 - 13 1136 171
          NM_053876.2 NP_446328.2 13 - 14 1019 171
          XM_006241116.2 XP_006241178.1 12 - 13 2326 171
          XM_008765198.1 XP_008763420.1 10 - 11 866 171
          GenBank mRNA AY724543.1 - 12 - 13 1123 171
          BC088317.1 - 8 - 9 647 7325202 171
          U32577.2 - 13 - 14 1019 171

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002292.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hnrnpm

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          alternative mRNA splicing, via spliceosome
          mRNA splicing, via spliceosome
          nucleotide binding
          RNA binding
          antigen binding
          protein binding
          protein domain specific binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P107794 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P016147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31830 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107794 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016147 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107794 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016147 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P107794 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P016147 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15645 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P548025 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700417046 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28910 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93140 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63746 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94544 - -
          Agilent Rat cDNA Microarray (G4105A) 700417046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10901150 - -
          Affymetrix Rat Gene 1.0 ST Array 10901151 - -
          Affymetrix Rat Gene 1.0 ST Array 10901146 - -
          Affymetrix Rat Gene 1.0 ST Array 10901144 - -
          Affymetrix Rat Gene 1.0 ST Array 10901148 - -
          Affymetrix Rat Gene 1.0 ST Array 10901143 - -
          Affymetrix Rat Gene 1.0 ST Array 10901149 - -
          Affymetrix Rat Gene 1.0 ST Array 10901147 - -
          Affymetrix Rat Gene 1.0 ST Array 10901145 - -
          Affymetrix Rat Gene 1.1 ST Array 10822083 - -
          Affymetrix Rat Gene 1.1 ST Array 10901142 - -
          Rat Genome 230 2.0 Array 1370919_at - -
          Rat Genome 230 2.0 Array 1370919_PM_at - -
          Rat Genome 230 2.0 Array 1372571_PM_at - -
          Rat Genome 230 2.0 Array 1372571_at - -
          Affymetrix HT Rat Focus Array 1370919_PM_at - -
          Affymetrix HT Rat Focus Array 1370919_at - -
          Affymetrix Rat Clariom S Assay PSR0700018335.rn.1 TC0700002292.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018340.rn.1 TC0700002292.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018339.rn.1 TC0700002292.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018337.rn.1 TC0700002292.rn.1 RTA TC
          Rat Expression Set 230 1370919_PM_at - -
          Rat Expression Set 230 1370919_at - -
          Affymetrix Rat Gene 1.0 ST Array 10901142 - -
          Rat Genome U34 Set U32577_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21656748 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349120 - -
          RatRef-12 Expression BeadChip ILMN_57541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline