Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02348092_s1
          See other OLR1696 GE Assays ›
          Gene Symbol:
          Olr1696
          Gene Name:
          olfactory receptor 1696
          Gene Aliases:
          SCR D-8
          Chromosome Location:
          Chr.20: 878460 - 879425 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02348092_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olr1696
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206850.1 NP_996732.1 1 830 161
          GenBank mRNA AF034896.1 - 1052 161

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olr1696

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394363 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133883 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49309 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82098 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006778 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006778 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006778 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19382 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29108 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39206 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37036 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830799 - -
          Affymetrix Rat Gene 1.0 ST Array 10830798 - -
          Affymetrix Rat Gene 1.0 ST Array 10830797 - -
          Affymetrix Rat Gene 1.0 ST Array 10830803 - -
          Affymetrix Rat Gene 1.0 ST Array 10830801 - -
          Affymetrix Rat Gene 1.1 ST Array 10830800 - -
          Affymetrix Rat Gene 1.1 ST Array 10830802 - -
          Affymetrix Rat Gene 1.1 ST Array 10830796 - -
          Rat Genome 230 2.0 Array 1370741_at - -
          Rat Genome 230 2.0 Array 1370741_PM_at - -
          Affymetrix HT Rat Focus Array 1370741_PM_at - -
          Affymetrix HT Rat Focus Array 1370741_at - -
          Affymetrix Rat Clariom S Assay PSR2000007637.rn.1 TC2000000952.rn.1 RTA TC
          Rat Expression Set 230 1370741_at - -
          Rat Expression Set 230 1370741_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830796 - -
          Affymetrix Rat Gene 1.0 ST Array 10830802 - -
          Affymetrix Rat Gene 1.0 ST Array 10830800 - -
          Rat Genome U34 Set AF034896_f_at - -
          Rat Genome U34 Set AF034900mRNA_i_at - -
          Rat Genome U34 Set AF034899_r_at - -
          Rat Genome U34 Set AF091563_i_at - -
          Rat Genome U34 Set AF034897_at - -
          Rat Genome U34 Set AF034899_i_at - -
          Rat Genome U34 Set AF034896_f_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370679 - -
          RatRef-12 Expression BeadChip ILMN_59415 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline