Search
Search

Vom1r19| Gene Symbol: Vom1r19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008954.1 | NP_001008954.1 | 1 | 832 | 113 | |
| GenBank mRNA | AY510367.1 | - | 1 | 832 | 113 | |
| AY510368.1 | - | 1 | 832 | 113 | ||
Set Membership: |
|