Search
Search

Vom1r92| Gene Symbol: Vom1r92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008968.1 | NP_001008968.1 | 1 | 131 | 154 | |
| XM_006236847.1 | XP_006236909.1 | 2 | 206 | 154 | ||
| GenBank mRNA | AY510279.1 | - | 1 | 131 | 154 | |
Set Membership: |
|