Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn02348762_s1
See other VOM1R31 GE Assays ›
Gene Symbol:
Vom1r31
Gene Name:
vomeronasal 1 receptor 31
Gene Aliases:
V1re12
Chromosome Location:
Chr.Un|NW_007906935: 24525 - 25457 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn02348762_s1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Vom1r31
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001008969.1
NP_001008969.1
715
136
GenBank mRNA
AY510343.1
-
715
136
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TCUn_KL568466v100000001.rn.1
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
G-protein coupled receptor
3' Most
Recommended Assay
Rattus norvegicus
Inventoried
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Vom1r31
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
pheromone receptor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_64_P135718
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P135718
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P135718
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_64_P135718
-
-
Agilent Rat Genome CGH Microarray 105A (G4436A)
94277
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Gene 1.0 ST Array
10718642
-
-
Affymetrix Rat Gene 1.1 ST Array
10718641
-
-
Affymetrix Rat Clariom S Assay
PSRUn_KL568466v100000001.rn.1
TCUn_KL568466v100000001.rn.1
RTA TC
Affymetrix Rat Gene 1.0 ST Array
10718641
-
-
Array Name
Probe IDs
Targets
Target Type
Illumina ratRef-12 v1.0 expression beadchip
ILMN_1359258
-
-
RatRef-12 Expression BeadChip
ILMN_52860
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline