Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02348805_m1
          See other LY49S6 GE Assays ›
          Gene Symbol:
          Ly49s6
          Gene Name:
          Ly49 stimulatory receptor 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 164429048 - 164453171 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02348805_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly49s6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009488.1 NP_001009488.1 3 - 4 326 147
          GenBank mRNA AY649837.1 - 3 - 4 326 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003882.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly49s6

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101005 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174966 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104129 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019170 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104129 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019170 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104129 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30185 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90813 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866147 - -
          Affymetrix Rat Gene 1.0 ST Array 10866150 - -
          Affymetrix Rat Gene 1.0 ST Array 10866171 - -
          Affymetrix Rat Gene 1.0 ST Array 10866177 - -
          Affymetrix Rat Gene 1.0 ST Array 10866148 - -
          Affymetrix Rat Gene 1.0 ST Array 10866151 - -
          Affymetrix Rat Gene 1.1 ST Array 10866146 - -
          Affymetrix Rat Gene 1.1 ST Array 10866199 - -
          Affymetrix Rat Clariom S Assay PSR0400027714.rn.1 TC0400003882.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027716.rn.1 TC0400003882.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027718.rn.1 TC0400003882.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027717.rn.1 TC0400003882.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10866146 - -
          Affymetrix Rat Gene 1.0 ST Array 10866167 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361534 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline