Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02348820_g1
          See other LY49S7 GE Assays ›
          Gene Symbol:
          Ly49s7
          Gene Name:
          Ly49 stimulatory receptor 7
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02348820_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly49s7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009494.1 NP_001009494.1 3 - 4 339 116
          GenBank mRNA AY649833.1 - 3 - 4 339 116
          AY649840.1 - 3 - 4 331 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003886.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly49s7

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P137001 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P118897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164754 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140142 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74442 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P118897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137001 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137001 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P118897 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P137001 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P118897 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P118897 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22105 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866230 - -
          Affymetrix Rat Gene 1.0 ST Array 10866229 - -
          Affymetrix Rat Gene 1.0 ST Array 10866227 - -
          Affymetrix Rat Gene 1.0 ST Array 10866226 - -
          Affymetrix Rat Gene 1.0 ST Array 10866217 - -
          Affymetrix Rat Gene 1.0 ST Array 10866231 - -
          Affymetrix Rat Gene 1.0 ST Array 10866232 - -
          Affymetrix Rat Gene 1.0 ST Array 10866224 - -
          Affymetrix Rat Gene 1.0 ST Array 10866228 - -
          Affymetrix Rat Gene 1.0 ST Array 10866225 - -
          Affymetrix Rat Gene 1.0 ST Array 10866223 - -
          Affymetrix Rat Gene 1.1 ST Array 10866222 - -
          Rat Genome 230 2.0 Array 1388282_PM_s_at - -
          Rat Genome 230 2.0 Array 1371272_at - -
          Rat Genome 230 2.0 Array 1388282_s_at - -
          Rat Genome 230 2.0 Array 1371272_PM_at - -
          Affymetrix HT Rat Focus Array 1371272_at - -
          Affymetrix HT Rat Focus Array 1371272_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400027749.rn.1 TC0400003886.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027750.rn.1 TC0400003886.rn.1 RTA TC
          Rat Expression Set 230 1388282_s_at - -
          Rat Expression Set 230 1388282_PM_s_at - -
          Rat Expression Set 230 1371272_PM_at - -
          Rat Expression Set 230 1371272_at - -
          Affymetrix Rat Gene 1.0 ST Array 10866215 - -
          Affymetrix Rat Gene 1.0 ST Array 10866222 - -
          Rat Genome U34 Set U56863_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374495 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline