Search
Search

Ly49si1| Gene Symbol: Ly49si1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009497.1 | NP_001009497.1 | 2 - 3 | 196 | 79 | |
| XM_008763312.1 | XP_008761534.1 | 2 - 3 | 447 | 79 | ||
| XM_008763313.1 | XP_008761535.1 | 3 - 4 | 477 | 79 | ||
| XM_008763314.1 | XP_008761536.1 | 3 - 4 | 271 | 79 | ||
| XM_008763315.1 | XP_008761537.1 | 2 - 3 | 447 | 79 | ||
| XM_008763316.1 | XP_008761538.1 | 2 - 3 | 447 | 79 | ||
| XM_008763319.1 | XP_008761541.1 | 2 - 3 | 447 | 79 | ||
| GenBank mRNA | AY659935.1 | - | 2 - 3 | 196 | 79 | |
Set Membership: |
|