Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02349020_m1
          See other KLRA2 GE Assays ›
          Gene Symbol:
          Klra2
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 2
          Gene Aliases:
          Ly49i6
          Chromosome Location:
          Chr.4: 165005722 - 165026566 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02349020_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009718.1 NP_001009718.1 6 - 7 794 154
          XM_008763403.1 XP_008761625.1 5 - 6 856 154
          GenBank mRNA AY651018.1 - 6 - 7 794 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003884.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra2

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400567 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48262 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019205 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93232 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866208 - -
          Affymetrix Rat Gene 1.0 ST Array 10866210 - -
          Affymetrix Rat Gene 1.0 ST Array 10866214 - -
          Affymetrix Rat Gene 1.0 ST Array 10866211 - -
          Affymetrix Rat Gene 1.0 ST Array 10866213 - -
          Affymetrix Rat Gene 1.0 ST Array 10866209 - -
          Affymetrix Rat Gene 1.0 ST Array 10866212 - -
          Affymetrix Rat Gene 1.1 ST Array 10866207 - -
          Affymetrix Rat Clariom S Assay PSR0400027729.rn.1 TC0400003884.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027730.rn.1 TC0400003884.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10866207 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349560 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline