Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02349944_m1
          See other MUG2 GE Assays ›
          Gene Symbol:
          Mug2
          Gene Name:
          murinoglobulin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 154215262 - 154282608 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02349944_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mug2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001002826.1 NP_001002826.1 3458 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001712.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mug2

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          endopeptidase inhibitor activity
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P536097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16763 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P536097 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P536097 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P536097 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P153947 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P536097 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53468 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89641 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57570 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99525 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858485 - -
          Affymetrix Rat Gene 1.0 ST Array 10858480 - -
          Affymetrix Rat Gene 1.0 ST Array 10858479 - -
          Affymetrix Rat Gene 1.0 ST Array 10858473 - -
          Affymetrix Rat Gene 1.0 ST Array 10858478 - -
          Affymetrix Rat Gene 1.0 ST Array 10858477 - -
          Affymetrix Rat Gene 1.0 ST Array 10858483 - -
          Affymetrix Rat Gene 1.0 ST Array 10858484 - -
          Affymetrix Rat Gene 1.0 ST Array 10858482 - -
          Affymetrix Rat Gene 1.0 ST Array 10858471 - -
          Affymetrix Rat Gene 1.0 ST Array 10858474 - -
          Affymetrix Rat Gene 1.0 ST Array 10858472 - -
          Affymetrix Rat Gene 1.0 ST Array 10858486 - -
          Affymetrix Rat Gene 1.0 ST Array 10858475 - -
          Affymetrix Rat Gene 1.0 ST Array 10858481 - -
          Affymetrix Rat Gene 1.1 ST Array 10858470 - -
          Rat Genome 230 2.0 Array 1370027_a_at - -
          Rat Genome 230 2.0 Array 1388229_PM_a_at - -
          Rat Genome 230 2.0 Array 1370027_PM_a_at - -
          Rat Genome 230 2.0 Array 1388229_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400012284.rn.1 TC0400001712.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012285.rn.1 TC0400001712.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012289.rn.1 TC0400001712.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012287.rn.1 TC0400001712.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012291.rn.1 TC0400001712.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012288.rn.1 TC0400001712.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012286.rn.1 TC0400001712.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012290.rn.1 TC0400001712.rn.1 RTA TC
          Rat Expression Set 230 1388229_a_at - -
          Rat Expression Set 230 1370027_PM_a_at - -
          Rat Expression Set 230 1388229_PM_a_at - -
          Rat Expression Set 230 1370027_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858470 - -
          Rat Genome U34 Set M22359mRNA_s_at - -
          Rat Genome U34 Set J03524_s_at - -
          Rat Genome U34 Set M22360_s_at - -
          Rat Genome U34 Set M22993cds_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline