Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02376848_s1
          See other POLE3 GE Assays ›
          Gene Symbol:
          Pole3
          Gene Name:
          polymerase (DNA directed), epsilon 3, accessory subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 78380813 - 78384047 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02376848_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pole3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007652.2 NP_001007653.1 4 2453 97
          GenBank mRNA BC083800.1 - 4 2453 7108773 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pole3

          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          histone H3 acetylation
          DNA biosynthetic process
          molecular_function
          DNA-directed DNA polymerase activity
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16804 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15212 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16804 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15212 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16804 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16804 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15212 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15212 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P472170 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16804 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877444 - -
          Affymetrix Rat Gene 1.0 ST Array 10905187 - -
          Affymetrix Rat Gene 1.0 ST Array 10877445 - -
          Affymetrix Rat Gene 1.1 ST Array 10905186 - -
          Affymetrix Rat Gene 1.1 ST Array 10877443 - -
          Rat Genome 230 2.0 Array 1383208_PM_at - -
          Rat Genome 230 2.0 Array 1394003_PM_at - -
          Rat Genome 230 2.0 Array 1388833_PM_at - -
          Rat Genome 230 2.0 Array 1394003_at - -
          Rat Genome 230 2.0 Array 1383208_at - -
          Rat Genome 230 2.0 Array 1376494_at - -
          Rat Genome 230 2.0 Array 1388833_at - -
          Rat Genome 230 2.0 Array 1376494_PM_at - -
          Affymetrix HT Rat Focus Array 1383208_at - -
          Affymetrix HT Rat Focus Array 1376494_at - -
          Affymetrix HT Rat Focus Array 1394003_PM_at - -
          Affymetrix HT Rat Focus Array 1388833_PM_at - -
          Affymetrix HT Rat Focus Array 1376494_PM_at - -
          Affymetrix HT Rat Focus Array 1394003_at - -
          Affymetrix HT Rat Focus Array 1383208_PM_at - -
          Affymetrix HT Rat Focus Array 1388833_at - -
          Affymetrix Rat Clariom S Assay PSR0500021564.rn.1 TC0500002878.rn.1 RTA TC
          Rat Expression Set 230 1376494_at - -
          Rat Expression Set 230 1388833_at - -
          Rat Expression Set 230 1388833_PM_at - -
          Rat Expression Set 230 1376494_PM_at - -
          Rat Expression Set 230 1383208_PM_at - -
          Rat Expression Set 230 1383208_at - -
          Rat Expression Set 230 1394003_at - -
          Rat Expression Set 230 1394003_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877443 - -
          Rat Genome U34 Set rc_AA892270_g_at - -
          Rat Genome U34 Set rc_AA892270_at - -
          Rat Genome U34 Set rc_AI045001_at - -
          Rat Genome U34 Set rc_AA818074_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21234448 - -
          ABI Rat Genome Survey Microarray 22073926 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364265 - -
          RatRef-12 Expression BeadChip ILMN_56130 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline