Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02376944_s1
          See other MRFAP1 GE Assays ›
          Gene Symbol:
          Mrfap1
          Gene Name:
          Morf4 family associated protein 1
          Gene Aliases:
          Pgr1, RGD1306635
          Chromosome Location:
          Chr.14: 78973384 - 78975161 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02376944_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrfap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009264.1 NP_001009264.1 2 1374 116
          GenBank mRNA BC088308.1 - 2 1374 7324285 116
          FQ228750.1 - 2 1394 116

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrfap1

          Gene Ontology Categories:

          Function(s) Process(es)

          mannose metabolic process
          alpha-mannosidase activity
          protein binding
          zinc ion binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078431 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P544569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P544569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078431 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P544569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078431 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078431 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P544569 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P226858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777434 - -
          Affymetrix Rat Gene 1.0 ST Array 10773327 - -
          Affymetrix Rat Gene 1.0 ST Array 10773330 - -
          Affymetrix Rat Gene 1.0 ST Array 10773329 - -
          Affymetrix Rat Gene 1.0 ST Array 10773328 - -
          Affymetrix Rat Gene 1.1 ST Array 10773298 - -
          Rat Genome 230 2.0 Array 1388311_PM_at - -
          Rat Genome 230 2.0 Array 1388311_at - -
          Affymetrix HT Rat Focus Array 1388311_at - -
          Affymetrix HT Rat Focus Array 1388311_PM_at - -
          Rat Expression Set 230 1388311_at - -
          Rat Expression Set 230 1388311_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773298 - -
          Rat Genome U34 Set rc_AA799550_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21910145 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline