Search
Search
| Gene Symbol: Lrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009717.1 | NP_001009717.1 | 2 | 1056 | 104 | |
| GenBank mRNA | BC088434.1 | - | 2 | 1056 | 7124833 | 104 |
| FQ220477.1 | - | 2 | 1066 | 104 | ||
Set Membership: |
|