Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02377062_g1
          See other HMGB1 GE Assays ›
          Gene Symbol:
          Hmgb1
          Gene Name:
          high mobility group box 1
          Gene Aliases:
          Ac2-008, Hmg1
          Chromosome Location:
          Chr.12: 7090927 - 7092190 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02377062_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hmgb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012963.2 NP_037095.1 2143 94
          GenBank mRNA BC081839.1 - 2143 94

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hmgb1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          eye development
          positive regulation of protein phosphorylation
          positive regulation of mesenchymal cell proliferation
          inflammatory response to antigenic stimulus
          base-excision repair, DNA ligation
          chromatin remodeling
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          chemotaxis
          nervous system development
          circadian rhythm
          negative regulation of DNA replication
          positive regulation of cell proliferation
          response to heat
          response to glucose
          positive regulation of cell death
          positive regulation of neuron projection development
          positive regulation of smooth muscle cell migration
          negative regulation of RNA polymerase II transcriptional preinitiation complex assembly
          lung development
          positive regulation of cell migration
          neuron projection development
          actin cytoskeleton reorganization
          DNA geometric change
          response to lipopolysaccharide
          response to insulin
          positive regulation of myeloid cell apoptotic process
          V(D)J recombination
          response to interferon-gamma
          response to drug
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of DNA binding
          positive regulation of myeloid cell differentiation
          positive regulation of myoblast differentiation
          positive regulation of glycogen catabolic process
          positive regulation of protein kinase activity
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          regulation of inflammatory response
          male-specific defense response to bacterium
          induction of positive chemotaxis
          response to glucocorticoid
          myoblast proliferation
          cellular response to interleukin-1
          four-way junction DNA binding
          open form four-way junction DNA binding
          crossed form four-way junction DNA binding
          DNA binding
          bent DNA binding
          chromatin binding
          double-stranded DNA binding
          single-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          cytokine activity
          protein binding
          5S rRNA binding
          transcription factor binding
          heparin binding
          DNA binding, bending
          calcium-dependent protein kinase regulator activity
          protein kinase activator activity
          peptide binding
          poly(A) RNA binding
          protein dimerization activity
          RAGE receptor binding
          glycolipid binding
          repressing transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P031945 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072803 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P299671 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P299671 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031945 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P299671 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031945 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P299671 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P031945 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072803 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P103832 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15527 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700543746 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702120630 - -
          Agilent Rat cDNA Microarray (G4105A) 702120630 - -
          Agilent Rat cDNA Microarray (G4105A) 700543746 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763431 - -
          Affymetrix Rat Gene 1.0 ST Array 10742834 - -
          Affymetrix Rat Gene 1.0 ST Array 10813356 - -
          Affymetrix Rat Gene 1.0 ST Array 10937359 - -
          Affymetrix Rat Gene 1.0 ST Array 10902248 - -
          Affymetrix Rat Gene 1.0 ST Array 10906454 - -
          Affymetrix Rat Gene 1.0 ST Array 10788032 - -
          Affymetrix Rat Gene 1.0 ST Array 10818697 - -
          Affymetrix Rat Gene 1.0 ST Array 10729504 - -
          Affymetrix Rat Gene 1.0 ST Array 10861907 - -
          Affymetrix Rat Gene 1.0 ST Array 10825180 - -
          Affymetrix Rat Gene 1.0 ST Array 10709778 - -
          Affymetrix Rat Gene 1.0 ST Array 10729506 - -
          Affymetrix Rat Gene 1.0 ST Array 10729503 - -
          Affymetrix Rat Gene 1.0 ST Array 10709777 - -
          Affymetrix Rat Gene 1.0 ST Array 10867798 - -
          Affymetrix Rat Gene 1.0 ST Array 10739594 - -
          Affymetrix Rat Gene 1.0 ST Array 10871285 - -
          Affymetrix Rat Gene 1.0 ST Array 10729505 - -
          Affymetrix Rat Gene 1.0 ST Array 10704917 - -
          Affymetrix Rat Gene 1.0 ST Array 10812016 - -
          Affymetrix Rat Gene 1.0 ST Array 10834137 - -
          Affymetrix Rat Gene 1.0 ST Array 10763432 - -
          Affymetrix Rat Gene 1.0 ST Array 10709776 - -
          Affymetrix Rat Gene 1.0 ST Array 10804451 - -
          Affymetrix Rat Gene 1.0 ST Array 10824883 - -
          Affymetrix Rat Gene 1.0 ST Array 10923209 - -
          Affymetrix Rat Gene 1.0 ST Array 10903226 - -
          Affymetrix Rat Gene 1.0 ST Array 10861906 - -
          Affymetrix Rat Gene 1.1 ST Array 10739593 - -
          Affymetrix Rat Gene 1.1 ST Array 10906453 - -
          Affymetrix Rat Gene 1.1 ST Array 10788031 - -
          Affymetrix Rat Gene 1.1 ST Array 10709775 - -
          Affymetrix Rat Gene 1.1 ST Array 10824882 - -
          Affymetrix Rat Gene 1.1 ST Array 10763430 - -
          Affymetrix Rat Gene 1.1 ST Array 10902247 - -
          Affymetrix Rat Gene 1.1 ST Array 10729502 - -
          Affymetrix Rat Gene 1.1 ST Array 10813355 - -
          Affymetrix Rat Gene 1.1 ST Array 10742833 - -
          Affymetrix Rat Gene 1.1 ST Array 10903225 - -
          Affymetrix Rat Gene 1.1 ST Array 10804449 - -
          Affymetrix Rat Gene 1.1 ST Array 10825179 - -
          Affymetrix Rat Gene 1.1 ST Array 10834136 - -
          Affymetrix Rat Gene 1.1 ST Array 10871284 - -
          Affymetrix Rat Gene 1.1 ST Array 10812015 - -
          Affymetrix Rat Gene 1.1 ST Array 10861905 - -
          Affymetrix Rat Gene 1.1 ST Array 10937358 - -
          Affymetrix Rat Gene 1.1 ST Array 10923208 - -
          Affymetrix Rat Gene 1.1 ST Array 10867797 - -
          Affymetrix Rat Gene 1.1 ST Array 10704916 - -
          Rat Genome 230 2.0 Array 1368042_PM_a_at - -
          Rat Genome 230 2.0 Array 1368042_a_at - -
          Affymetrix HT Rat Focus Array 1368042_a_at - -
          Affymetrix HT Rat Focus Array 1388783_at - -
          Affymetrix HT Rat Focus Array 1388783_PM_at - -
          Affymetrix HT Rat Focus Array 1368042_PM_a_at - -
          Rat Expression Set 230 1368042_a_at - -
          Rat Expression Set 230 1368042_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709775 - -
          Affymetrix Rat Gene 1.0 ST Array 10861905 - -
          Affymetrix Rat Gene 1.0 ST Array 10867797 - -
          Affymetrix Rat Gene 1.0 ST Array 10788031 - -
          Affymetrix Rat Gene 1.0 ST Array 10812015 - -
          Affymetrix Rat Gene 1.0 ST Array 10903225 - -
          Affymetrix Rat Gene 1.0 ST Array 10704916 - -
          Affymetrix Rat Gene 1.0 ST Array 10739593 - -
          Affymetrix Rat Gene 1.0 ST Array 10923208 - -
          Affymetrix Rat Gene 1.0 ST Array 10906453 - -
          Affymetrix Rat Gene 1.0 ST Array 10937358 - -
          Affymetrix Rat Gene 1.0 ST Array 10742833 - -
          Affymetrix Rat Gene 1.0 ST Array 10871284 - -
          Affymetrix Rat Gene 1.0 ST Array 10729502 - -
          Rat Genome U34 Set M64986_at - -
          Rat Genome U34 Set M64986_g_at - -
          Rat Genome U34 Set rc_AI045885_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21557521 - -
          ABI Rat Genome Survey Microarray 21707264 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359535 - -
          RatRef-12 Expression BeadChip ILMN_53698 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline