Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02377325_s1
          See other SH2B3 GE Assays ›
          Gene Symbol:
          Sh2b3
          Gene Name:
          SH2B adaptor protein 3
          Gene Aliases:
          Lnk
          Chromosome Location:
          Chr.12: 40261990 - 40265757 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02377325_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sh2b3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031621.1 NP_113809.1 2 960 115
          GenBank mRNA U24655.1 - 2 960 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sh2b3

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          hemopoiesis
          cell differentiation
          embryonic hemopoiesis
          intracellular signal transduction
          protein complex binding
          phosphate ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P034435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22691 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P034435 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P034435 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P034435 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P401536 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700933956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100469 - -
          Agilent Rat cDNA Microarray (G4105A) 700933956 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10758650 - -
          Affymetrix Rat Gene 1.0 ST Array 10758647 - -
          Affymetrix Rat Gene 1.0 ST Array 10758654 - -
          Affymetrix Rat Gene 1.0 ST Array 10758648 - -
          Affymetrix Rat Gene 1.0 ST Array 10758649 - -
          Affymetrix Rat Gene 1.0 ST Array 10758652 - -
          Affymetrix Rat Gene 1.0 ST Array 10758657 - -
          Affymetrix Rat Gene 1.0 ST Array 10758655 - -
          Affymetrix Rat Gene 1.0 ST Array 10758653 - -
          Affymetrix Rat Gene 1.0 ST Array 10758656 - -
          Affymetrix Rat Gene 1.0 ST Array 10758651 - -
          Affymetrix Rat Gene 1.1 ST Array 10758645 - -
          Rat Genome 230 2.0 Array 1367723_PM_a_at - -
          Rat Genome 230 2.0 Array 1367723_a_at - -
          Affymetrix HT Rat Focus Array 1367723_a_at - -
          Affymetrix HT Rat Focus Array 1367723_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1200005917.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005918.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005913.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005914.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005920.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005921.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005922.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005915.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005916.rn.1 TC1200000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200005923.rn.1 TC1200000661.rn.1 RTA TC
          Rat Expression Set 230 1367723_PM_a_at - -
          Rat Expression Set 230 1367723_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10758645 - -
          Rat Genome U34 Set U24652_at - -
          Rat Genome U34 Set rc_AA943555_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376327 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline