Search
Search
| Gene Symbol: Kb23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008813.1 | NP_001008813.1 | 2 - 3 | 562 | 74 | |
| XM_006242350.2 | XP_006242412.1 | 2 - 3 | 607 | 74 | ||
| GenBank mRNA | AB355639.1 | - | 2 - 3 | 562 | 74 | |
Set Membership: |
|