Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02395312_m1
          See other CDH20 GE Assays ›
          Gene Symbol:
          Cdh20
          Gene Name:
          cadherin 20
          Gene Aliases:
          Cad20
          Chromosome Location:
          Chr.13: 24846480 - 24913440 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02395312_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdh20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012748.1 NP_001012766.1 10 - 11 1902 60
          GenBank mRNA AB121033.1 - 10 - 11 1902 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000063.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdh20

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P033969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468966 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81832 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134799 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033969 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P033969 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57400 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19501 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763231 - -
          Affymetrix Rat Gene 1.0 ST Array 10763228 - -
          Affymetrix Rat Gene 1.0 ST Array 10763232 - -
          Affymetrix Rat Gene 1.0 ST Array 10763234 - -
          Affymetrix Rat Gene 1.0 ST Array 10763236 - -
          Affymetrix Rat Gene 1.0 ST Array 10763226 - -
          Affymetrix Rat Gene 1.0 ST Array 10763235 - -
          Affymetrix Rat Gene 1.0 ST Array 10763227 - -
          Affymetrix Rat Gene 1.0 ST Array 10763229 - -
          Affymetrix Rat Gene 1.0 ST Array 10763233 - -
          Affymetrix Rat Gene 1.0 ST Array 10763230 - -
          Affymetrix Rat Gene 1.1 ST Array 10763225 - -
          Rat Genome 230 2.0 Array 1393897_PM_at - -
          Rat Genome 230 2.0 Array 1393897_at - -
          Affymetrix HT Rat Focus Array 1393897_PM_at - -
          Affymetrix HT Rat Focus Array 1393897_at - -
          Affymetrix Rat Clariom S Assay PSR1300000234.rn.1 TC1300000063.rn.1 RTA TC
          Rat Expression Set 230 1393897_PM_at - -
          Rat Expression Set 230 1393897_at - -
          Affymetrix Rat Gene 1.0 ST Array 10763225 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355171 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline