Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02395603_s1
          See other MTERFD3 GE Assays ›
          Gene Symbol:
          Mterfd3
          Gene Name:
          MTERF domain containing 3
          Gene Aliases:
          RGD1311836, mTERF2
          Chromosome Location:
          Chr.7: 24638208 - 24648263 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02395603_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mterfd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014265.2 NP_001014287.1 4 1472 95
          GenBank mRNA BC083741.1 - 1 1337 7191829 95
          BC098693.1 - 4 1472 7386232 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mterfd3

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          double-stranded DNA binding
          protein homodimerization activity
          transcription regulatory region DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P456172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P456172 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P456172 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P456172 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P456172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95236 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894541 - -
          Affymetrix Rat Gene 1.0 ST Array 10894543 - -
          Affymetrix Rat Gene 1.0 ST Array 10894542 - -
          Affymetrix Rat Gene 1.0 ST Array 10894545 - -
          Affymetrix Rat Gene 1.0 ST Array 10894544 - -
          Affymetrix Rat Gene 1.1 ST Array 10894540 - -
          Rat Genome 230 2.0 Array 1378034_at - -
          Rat Genome 230 2.0 Array 1378034_PM_at - -
          Affymetrix HT Rat Focus Array 1378034_PM_at - -
          Affymetrix HT Rat Focus Array 1378034_at - -
          Affymetrix Rat Clariom S Assay PSR0700003360.rn.1 TC0700000393.rn.1 RTA TC
          Rat Expression Set 230 1378034_PM_at - -
          Rat Expression Set 230 1378034_at - -
          Affymetrix Rat Gene 1.0 ST Array 10894540 - -
          Rat Genome U34 Set rc_AA850498_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22317957 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370145 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline