Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02395903_m1
          See other BRI3BP GE Assays ›
          Gene Symbol:
          Bri3bp
          Gene Name:
          Bri3 binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 36576018 - 36587839 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02395903_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bri3bp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017487.1 NP_001017487.1 2 - 3 348 105
          GenBank mRNA BC085334.1 - 2 - 3 348 7301029 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001514.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bri3bp

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P607722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P607722 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P607722 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P607722 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P607722 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P728719 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16196 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24563 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761548 - -
          Affymetrix Rat Gene 1.0 ST Array 10761547 - -
          Affymetrix Rat Gene 1.0 ST Array 10761546 - -
          Affymetrix Rat Gene 1.1 ST Array 10761545 - -
          Rat Genome 230 2.0 Array 1392839_at - -
          Rat Genome 230 2.0 Array 1392839_PM_at - -
          Affymetrix HT Rat Focus Array 1392839_PM_at - -
          Affymetrix HT Rat Focus Array 1392839_at - -
          Affymetrix Rat Clariom S Assay PSR1200012644.rn.1 TC1200001514.rn.1 RTA TC
          Rat Expression Set 230 1392839_at - -
          Rat Expression Set 230 1392839_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761545 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21982323 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356542 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline