Search
Search
| Gene Symbol: Spib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024286.1 | NP_001019457.1 | 4 - 5 | 370 | 62 | |
| XM_006229111.2 | XP_006229173.1 | 3 - 4 | 1119 | 62 | ||
| XM_006229112.2 | XP_006229174.1 | 4 - 5 | 439 | 62 | ||
| XM_006229113.2 | XP_006229175.1 | 3 - 4 | 1119 | 62 | ||
| XM_006229114.2 | XP_006229176.1 | 3 - 4 | 1064 | 62 | ||
| XM_006229115.2 | XP_006229177.1 | 4 - 5 | 384 | 62 | ||
| XM_008759404.1 | XP_008757626.1 | 3 - 4 | 1067 | 62 | ||
| XM_008759405.1 | XP_008757627.1 | 3 - 4 | 420 | 62 | ||
| GenBank mRNA | BC089881.1 | - | 4 - 5 | 370 | 7368473 | 62 |
Set Membership: |
|