Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02396213_m1
          See other LOC499886 GE Assays ›
          Gene Symbol:
          LOC499886
          Gene Name:
          similar to hypothetical protein 4933411G11
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 121128312 - 121135720 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02396213_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC499886
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024312.1 NP_001019483.1 6 - 7 774 69
          XM_008762191.1 XP_008760413.1 6 - 7 830 69
          XM_008762192.1 XP_008760414.1 6 - 7 830 69
          GenBank mRNA BC086511.1 - 6 - 7 774 7132989 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001678.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC499886

          Gene Ontology Categories:

          Process(es)

          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11804 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34619 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839668 - -
          Affymetrix Rat Gene 1.0 ST Array 10839673 - -
          Affymetrix Rat Gene 1.0 ST Array 10839667 - -
          Affymetrix Rat Gene 1.0 ST Array 10839671 - -
          Affymetrix Rat Gene 1.0 ST Array 10839669 - -
          Affymetrix Rat Gene 1.0 ST Array 10839663 - -
          Affymetrix Rat Gene 1.0 ST Array 10839665 - -
          Affymetrix Rat Gene 1.0 ST Array 10839670 - -
          Affymetrix Rat Gene 1.0 ST Array 10839666 - -
          Affymetrix Rat Gene 1.0 ST Array 10839664 - -
          Affymetrix Rat Gene 1.0 ST Array 10839672 - -
          Affymetrix Rat Gene 1.1 ST Array 10839662 - -
          Affymetrix Rat Clariom S Assay PSR0300011949.rn.1 TC0300001678.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10839662 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366943 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline