Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02397252_gH
          See other LOC259245 GE Assays ›
          Gene Symbol:
          LOC259245
          Gene Name:
          alpha-2u globulin PGCL5
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02397252_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC259245
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147213.1 NP_671746.1 5 - 6 561 168
          GenBank mRNA AB039826.1 - 5 - 6 561 168

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002863.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          transporter activity
          pheromone binding
          small molecule binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P387145 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P387145 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P387145 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123338 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P387145 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123338 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P220403 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P387145 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P334779 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877311 - -
          Affymetrix Rat Gene 1.0 ST Array 10877353 - -
          Affymetrix Rat Gene 1.0 ST Array 10877293 - -
          Affymetrix Rat Gene 1.0 ST Array 10877322 - -
          Affymetrix Rat Gene 1.0 ST Array 10877318 - -
          Affymetrix Rat Gene 1.0 ST Array 10877350 - -
          Affymetrix Rat Gene 1.0 ST Array 10877345 - -
          Affymetrix Rat Gene 1.0 ST Array 10877351 - -
          Affymetrix Rat Gene 1.0 ST Array 10877323 - -
          Affymetrix Rat Gene 1.0 ST Array 10877291 - -
          Affymetrix Rat Gene 1.0 ST Array 10877310 - -
          Affymetrix Rat Gene 1.0 ST Array 10877282 - -
          Affymetrix Rat Gene 1.0 ST Array 10877281 - -
          Affymetrix Rat Gene 1.0 ST Array 10877364 - -
          Affymetrix Rat Gene 1.0 ST Array 10877285 - -
          Affymetrix Rat Gene 1.0 ST Array 10877239 - -
          Affymetrix Rat Gene 1.0 ST Array 10877248 - -
          Affymetrix Rat Gene 1.0 ST Array 10877305 - -
          Affymetrix Rat Gene 1.0 ST Array 10877298 - -
          Affymetrix Rat Gene 1.0 ST Array 10877288 - -
          Affymetrix Rat Gene 1.0 ST Array 10877321 - -
          Affymetrix Rat Gene 1.0 ST Array 10877319 - -
          Affymetrix Rat Gene 1.0 ST Array 10877347 - -
          Affymetrix Rat Gene 1.0 ST Array 10877232 - -
          Affymetrix Rat Gene 1.0 ST Array 10877320 - -
          Affymetrix Rat Gene 1.0 ST Array 10877283 - -
          Affymetrix Rat Gene 1.0 ST Array 10877279 - -
          Affymetrix Rat Gene 1.0 ST Array 10877258 - -
          Affymetrix Rat Gene 1.0 ST Array 10877316 - -
          Affymetrix Rat Gene 1.0 ST Array 10877238 - -
          Affymetrix Rat Gene 1.0 ST Array 10877284 - -
          Affymetrix Rat Gene 1.0 ST Array 10877361 - -
          Affymetrix Rat Gene 1.0 ST Array 10877249 - -
          Affymetrix Rat Gene 1.0 ST Array 10877303 - -
          Affymetrix Rat Gene 1.1 ST Array 10877313 - -
          Affymetrix Rat Gene 1.1 ST Array 10877307 - -
          Affymetrix Rat Gene 1.1 ST Array 10877278 - -
          Rat Genome 230 2.0 Array 1388031_PM_x_at - -
          Rat Genome 230 2.0 Array 1388031_x_at - -
          Affymetrix HT Rat Focus Array 1388031_x_at - -
          Affymetrix HT Rat Focus Array 1388031_PM_x_at - -
          Affymetrix Rat Clariom S Assay PSR0500021356.rn.1 TC0500002863.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021360.rn.1 TC0500002863.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021363.rn.1 TC0500002863.rn.1 RTA TC
          Rat Expression Set 230 1388031_x_at - -
          Rat Expression Set 230 1388031_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877307 - -
          Affymetrix Rat Gene 1.0 ST Array 10877313 - -
          Affymetrix Rat Gene 1.0 ST Array 10877286 - -
          Affymetrix Rat Gene 1.0 ST Array 10877278 - -
          Affymetrix Rat Gene 1.0 ST Array 10877339 - -
          Affymetrix Rat Gene 1.0 ST Array 10877295 - -
          Affymetrix Rat Gene 1.0 ST Array 10877251 - -
          Affymetrix Rat Gene 1.0 ST Array 10877229 - -
          Rat Genome U34 Set M24109mRNA_at - -
          Rat Genome U34 Set M24108mRNA_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21580420 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350838 - -
          RatRef-12 Expression BeadChip ILMN_52583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline