Search
Search
| Gene Symbol: P2ry14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133577.1 | NP_598261.1 | 1564 | 74 | ||
| XM_006232396.2 | XP_006232458.1 | 2 | 8623 | 74 | ||
| XM_008760984.1 | XP_008759206.1 | 2 | 6886 | 74 | ||
| XM_008760985.1 | XP_008759207.1 | 2 | 4465 | 74 | ||
| XM_008760986.1 | XP_008759208.1 | 2 | 4446 | 74 | ||
| GenBank mRNA | BC083545.1 | - | 2 | 1645 | 7191572 | 74 |
| U76206.1 | - | 2 | 1564 | 74 | ||
Set Membership: |
|