Search
Search

Fam110a| Gene Symbol: Fam110a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014050.2 | NP_001014072.2 | 2 | 986 | 78 | |
| NM_001244768.1 | NP_001231697.1 | 2 | 867 | 78 | ||
| NM_001244769.1 | NP_001231698.1 | 2 | 939 | 78 | ||
| NM_001244770.1 | NP_001231699.1 | 2 | 786 | 78 | ||
| XM_006235231.2 | XP_006235293.1 | 2 | 1239 | 78 | ||
| XM_006235234.2 | XP_006235296.1 | 3 | 974 | 78 | ||
| GenBank mRNA | BC079251.1 | - | 2 | 986 | 7133086 | 78 |
| CB766675.1 | - | 1 | 99 | 78 | ||
| CB806736.1 | - | 1 | 276 | 78 | ||
| DY318143.1 | - | 1 | 667 | 78 | ||
| JF912497.1 | - | 1 | 597 | 78 | ||
Set Membership: |
|