Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02533143_s1
          See other MGC105567 GE Assays ›
          Gene Symbol:
          MGC105567
          Gene Name:
          similar to cDNA sequence BC023105
          Gene Aliases:
          Ifgga5
          Chromosome Location:
          Chr.18: 55685613 - 55704910 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02533143_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MGC105567
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024284.1 NP_001019455.1 2 1066 103
          GenBank mRNA BC093614.1 - 2 1066 7309663 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MGC105567

          Gene Ontology Categories:

          Process(es)

          GTP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616055 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164764 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164764 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164764 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64002 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64767 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10801981 - -
          Affymetrix Rat Gene 1.0 ST Array 10801982 - -
          Affymetrix Rat Gene 1.1 ST Array 10801980 - -
          Affymetrix Rat Clariom S Assay PSR1800004399.rn.1 TC1800000610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004398.rn.1 TC1800000610.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10801980 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0