Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02534308_s1
          See other RNF146 GE Assays ›
          Gene Symbol:
          Rnf146
          Gene Name:
          ring finger protein 146
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 30856930 - 30873814 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02534308_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf146
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012060.2 NP_001012060.2 3 1062 95
          XM_006227744.2 XP_006227806.1 3 1119 95
          XM_006227745.2 XP_006227807.1 4 1167 95
          GenBank mRNA BC083675.1 - 3 1062 7135728 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf146

          Gene Ontology Categories:

          Function(s) Process(es)

          Wnt signaling pathway
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          protein autoubiquitination
          protein K48-linked ubiquitination
          positive regulation of canonical Wnt signaling pathway
          ubiquitin-protein transferase activity
          zinc ion binding
          ligase activity
          poly-ADP-D-ribose binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P083094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95115 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12818 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083094 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083094 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P083094 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P365598 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702426 - -
          Affymetrix Rat Gene 1.0 ST Array 10702427 - -
          Affymetrix Rat Gene 1.0 ST Array 10702425 - -
          Affymetrix Rat Gene 1.1 ST Array 10702424 - -
          Rat Genome 230 2.0 Array 1371778_at - -
          Rat Genome 230 2.0 Array 1371778_PM_at - -
          Affymetrix HT Rat Focus Array 1371778_at - -
          Affymetrix HT Rat Focus Array 1371778_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100001779.rn.1 TC0100000313.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001776.rn.1 TC0100000313.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001777.rn.1 TC0100000313.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001778.rn.1 TC0100000313.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001781.rn.1 TC0100000313.rn.1 RTA TC
          Rat Expression Set 230 1371778_at - -
          Rat Expression Set 230 1371778_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702424 - -
          Rat Genome U34 Set rc_AI177765_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22253437 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376209 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline