Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02534324_s1
          See other SLC35G2 GE Assays ›
          Gene Symbol:
          Slc35g2
          Gene Name:
          solute carrier family 35, member G2
          Gene Aliases:
          Tmem22
          Chromosome Location:
          Chr.8: 108854144 - 108881317 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02534324_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc35g2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012126.1 NP_001012126.1 2 1308 147
          XM_006243630.2 XP_006243692.1 2 1476 147
          GenBank mRNA BC088758.1 - 2 1308 7315776 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc35g2

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P048245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50782 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73321 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82677 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130060 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048245 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048245 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P048245 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13457 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9709 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98771 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919553 - -
          Affymetrix Rat Gene 1.0 ST Array 10919554 - -
          Affymetrix Rat Gene 1.1 ST Array 10919552 - -
          Rat Genome 230 2.0 Array 1392045_PM_at - -
          Rat Genome 230 2.0 Array 1392045_at - -
          Affymetrix HT Rat Focus Array 1392045_PM_at - -
          Affymetrix HT Rat Focus Array 1392045_at - -
          Affymetrix Rat Clariom S Assay PSR0800025830.rn.1 TC0800003242.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800025829.rn.1 TC0800003242.rn.1 RTA TC
          Rat Expression Set 230 1392045_PM_at - -
          Rat Expression Set 230 1392045_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919552 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21064292 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376675 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline