Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02534450_sH
          See other LY49S5 GE Assays ›
          Gene Symbol:
          Ly49s5
          Gene Name:
          Ly49 stimulatory receptor 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 164284910 - 164306500 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02534450_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly49s5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012749.1 NP_001012767.1 7 1234 134
          GenBank mRNA AY649832.1 - 7 1234 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003880.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly49s5

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46010 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032777 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032777 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032777 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58966 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866171 - -
          Affymetrix Rat Gene 1.0 ST Array 10866168 - -
          Affymetrix Rat Gene 1.0 ST Array 10866158 - -
          Affymetrix Rat Gene 1.0 ST Array 10866155 - -
          Affymetrix Rat Gene 1.0 ST Array 10866177 - -
          Affymetrix Rat Gene 1.0 ST Array 10866159 - -
          Affymetrix Rat Gene 1.0 ST Array 10866154 - -
          Affymetrix Rat Gene 1.1 ST Array 10866167 - -
          Affymetrix Rat Gene 1.1 ST Array 10866153 - -
          Affymetrix Rat Clariom S Assay PSR0400027691.rn.1 TC0400003880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027692.rn.1 TC0400003880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027693.rn.1 TC0400003880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027690.rn.1 TC0400003880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027694.rn.1 TC0400003880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027688.rn.1 TC0400003880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027689.rn.1 TC0400003880.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10866153 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline