Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02585812_m1
          See other KPNA5 GE Assays ›
          Gene Symbol:
          Kpna5
          Gene Name:
          karyopherin alpha 5 (importin alpha 6)
          Gene Aliases:
          Ipoa6
          Chromosome Location:
          Chr.20: 32509573 - 32573591 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02585812_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kpna5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025113.1 NP_001020284.1 12 - 13 1421 82
          XM_006256442.2 XP_006256504.1 13 - 14 1601 82
          XM_008772879.1 XP_008771101.1 11 - 12 1276 82
          GenBank mRNA AY779029.1 - 12 - 13 1421 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000706.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kpna5

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus
          protein transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P021795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136813 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52755 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021795 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021795 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P021795 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 806 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68443 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22163 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830101 - -
          Affymetrix Rat Gene 1.0 ST Array 10830099 - -
          Affymetrix Rat Gene 1.0 ST Array 10830094 - -
          Affymetrix Rat Gene 1.0 ST Array 10830090 - -
          Affymetrix Rat Gene 1.0 ST Array 10830091 - -
          Affymetrix Rat Gene 1.0 ST Array 10830097 - -
          Affymetrix Rat Gene 1.0 ST Array 10830089 - -
          Affymetrix Rat Gene 1.0 ST Array 10830095 - -
          Affymetrix Rat Gene 1.0 ST Array 10830096 - -
          Affymetrix Rat Gene 1.0 ST Array 10830092 - -
          Affymetrix Rat Gene 1.0 ST Array 10830093 - -
          Affymetrix Rat Gene 1.0 ST Array 10830098 - -
          Affymetrix Rat Gene 1.0 ST Array 10830100 - -
          Affymetrix Rat Gene 1.1 ST Array 10830088 - -
          Affymetrix Rat Clariom S Assay PSR2000006132.rn.1 TC2000000706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006131.rn.1 TC2000000706.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10830088 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374811 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline