Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02585827_g1
          See other RGD1561328 GE Assays ›
          Gene Symbol:
          RGD1561328
          Gene Name:
          cysteine and histidine rich 1-like
          Gene Aliases:
          Cyhr1
          Chromosome Location:
          Chr.7: 117720909 - 117722359 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02585827_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1561328
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025122.2 NP_001020293.2 1 - 2 491 102
          GenBank mRNA BC091178.1 - 1 - 2 491 7325408 102
          DV715431.1 - 1 - 2 218 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700004061.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGD1561328

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P122735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763152 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161622 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122735 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122735 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P122735 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31185 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905083 - -
          Affymetrix Rat Gene 1.0 ST Array 10905081 - -
          Affymetrix Rat Gene 1.0 ST Array 10905082 - -
          Affymetrix Rat Gene 1.0 ST Array 10905085 - -
          Affymetrix Rat Gene 1.0 ST Array 10905086 - -
          Affymetrix Rat Gene 1.0 ST Array 10905080 - -
          Affymetrix Rat Gene 1.0 ST Array 10905084 - -
          Affymetrix Rat Gene 1.1 ST Array 10905078 - -
          Rat Genome 230 2.0 Array 1377176_at - -
          Rat Genome 230 2.0 Array 1377738_a_at - -
          Rat Genome 230 2.0 Array 1377176_PM_at - -
          Rat Genome 230 2.0 Array 1377738_PM_a_at - -
          Rat Genome 230 2.0 Array 1382892_PM_a_at - -
          Rat Genome 230 2.0 Array 1382892_a_at - -
          Affymetrix HT Rat Focus Array 1377176_at - -
          Affymetrix HT Rat Focus Array 1377738_a_at - -
          Affymetrix HT Rat Focus Array 1377738_PM_a_at - -
          Affymetrix HT Rat Focus Array 1377176_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700033437.rn.1 TC0700004061.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033438.rn.1 TC0700004061.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033439.rn.1 TC0700004061.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033436.rn.1 TC0700004061.rn.1 RTA TC
          Rat Expression Set 230 1382892_PM_a_at - -
          Rat Expression Set 230 1377176_at - -
          Rat Expression Set 230 1382892_a_at - -
          Rat Expression Set 230 1377176_PM_at - -
          Rat Expression Set 230 1377738_a_at - -
          Rat Expression Set 230 1377738_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905078 - -
          Rat Genome U34 Set rc_AA799721_g_at - -
          Rat Genome U34 Set rc_AA799721_at - -
          Rat Genome U34 Set rc_AI045584_at - -
          Rat Genome U34 Set rc_AI010295_at - -
          Rat Genome U34 Set rc_AI171375_at - -
          Rat Genome U34 Set rc_AI172358_s_at - -
          Rat Genome U34 Set rc_AI176855_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365300 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline