Search
Search
| Gene Symbol: Hyou1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034028.1 | NP_001029200.1 | 8 - 9 | 839 | 76 | |
| NM_138867.2 | NP_620222.2 | 8 - 9 | 991 | 76 | ||
| XM_006242884.2 | XP_006242946.1 | 8 - 9 | 936 | 76 | ||
| XM_006242885.2 | XP_006242947.1 | 8 - 9 | 872 | 76 | ||
| XM_008766143.1 | XP_008764365.1 | 8 - 9 | 918 | 76 | ||
| GenBank mRNA | BC065310.1 | - | 8 - 9 | 882 | 5621638 | 76 |
| U41853.1 | - | 8 - 9 | 991 | 76 | ||
Set Membership: |
|