Search
Search

Vom1r101| Gene Symbol: Vom1r101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173112.2 | NP_775135.2 | 1 | 884 | 86 | |
| GenBank mRNA | AY510287.1 | - | 1 | 884 | 86 | |
| U36786.1 | - | 1 | 802 | 86 | ||
Set Membership: |
|