Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02748654_g1
          See other NDUFB4 GE Assays ›
          Gene Symbol:
          Ndufb4
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 65960277 - 65966899 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02748654_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufb4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037338.1 NP_001032415.1 1 - 2 193 81
          GenBank mRNA CB732095.1 - 1 - 2 193 81
          DQ255901.1 - 1 - 2 46 81
          FQ217067.1 - 1 - 2 200 81

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufb4

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to oxidative stress
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078769 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079483 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P073625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137726 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078769 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073625 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078769 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073625 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078769 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P073625 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P655706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751351 - -
          Affymetrix Rat Gene 1.0 ST Array 10839231 - -
          Affymetrix Rat Gene 1.0 ST Array 10751349 - -
          Affymetrix Rat Gene 1.0 ST Array 10751350 - -
          Affymetrix Rat Gene 1.0 ST Array 10839230 - -
          Affymetrix Rat Gene 1.1 ST Array 10839229 - -
          Affymetrix Rat Gene 1.1 ST Array 10751348 - -
          Rat Genome 230 2.0 Array 1371371_at - -
          Rat Genome 230 2.0 Array 1371371_PM_at - -
          Affymetrix HT Rat Focus Array 1371371_at - -
          Affymetrix HT Rat Focus Array 1371371_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100003531.rn.1 TC1100000662.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003528.rn.1 TC1100000662.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003530.rn.1 TC1100000662.rn.1 RTA TC
          Rat Expression Set 230 1371371_at - -
          Rat Expression Set 230 1371371_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751348 - -
          Affymetrix Rat Gene 1.0 ST Array 10839229 - -
          Rat Genome U34 Set rc_AA859957_at - -
          Rat Genome U34 Set rc_AA859957_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21351461 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374243 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351573 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline