Search
Search
| Gene Symbol: Defb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037509.1 | NP_001032598.1 | 1 - 2 | 56 | 90 | |
| XM_006253374.2 | XP_006253436.1 | 1 - 2 | 164 | 90 | ||
| GenBank mRNA | AY621341.1 | - | 1 - 2 | 56 | 90 | |
Set Membership: |
|