Search
Search
| Gene Symbol: Defb30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037531.1 | NP_001032620.1 | 1 - 2 | 52 | 112 | |
| XM_008770776.1 | XP_008768998.1 | 1 - 2 | 96 | 112 | ||
| GenBank mRNA | AY600146.1 | - | 1 - 2 | 102 | 112 | |
| AY621361.1 | - | 1 - 2 | 52 | 112 | ||
Set Membership: |
|