Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02758772_s1
          See other AGTR1A GE Assays ›
          Gene Symbol:
          Agtr1a
          Gene Name:
          angiotensin II receptor, type 1a
          Gene Aliases:
          AT1, AT1A, AT1R, Agtr1
          Chromosome Location:
          Chr.17: 35906084 - 35958132 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02758772_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Agtr1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030985.4 NP_112247.2 3 1235 112
          XM_006253882.2 XP_006253944.1 3 1274 112
          XM_008771593.1 XP_008769815.1 2 1191 112
          XM_008771594.1 XP_008769816.1 2 1207 112
          GenBank mRNA BC078810.1 - 3 1235 7126918 112
          M74054.1 - 3 1230 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Agtr1a

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          kidney development
          positive regulation of receptor recycling
          regulation of systemic arterial blood pressure by circulatory renin-angiotensin
          renin secretion into blood stream
          renin-angiotensin regulation of aldosterone production
          regulation of renal output by angiotensin
          brain renin-angiotensin system
          regulation of pH
          inflammatory response
          G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          Rho protein signal transduction
          heart development
          aging
          regulation of blood pressure
          positive regulation of cell proliferation
          positive regulation of cholesterol esterification
          regulation of vasoconstriction
          calcium-mediated signaling
          positive regulation of cellular protein metabolic process
          positive regulation of phospholipase A2 activity
          positive regulation of superoxide anion generation
          regulation of smooth muscle cell apoptotic process
          negative regulation of smooth muscle cell apoptotic process
          angiotensin-activated signaling pathway
          vasoconstriction
          dopamine biosynthetic process
          drinking behavior
          activation of Janus kinase activity
          negative regulation of neuron apoptotic process
          response to estrogen
          positive regulation of blood pressure
          positive regulation of cytokine secretion
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          cell chemotaxis
          cellular response to dexamethasone stimulus
          phospholipase C-activating angiotensin-activated signaling pathway
          positive regulation of branching involved in ureteric bud morphogenesis
          angiotensin type I receptor activity
          G-protein alpha-subunit binding
          angiotensin type II receptor activity
          protein binding
          peptide hormone binding
          protein kinase binding
          bradykinin receptor binding
          D1 dopamine receptor binding
          protein self-association
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P486203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146420 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P486203 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P486203 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P486203 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P476936 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P486203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31396 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80749 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38160 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12846 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798197 - -
          Affymetrix Rat Gene 1.0 ST Array 10798196 - -
          Affymetrix Rat Gene 1.0 ST Array 10798198 - -
          Affymetrix Rat Gene 1.0 ST Array 10798195 - -
          Affymetrix Rat Gene 1.1 ST Array 10798194 - -
          Rat Genome 230 2.0 Array 1369291_PM_at - -
          Rat Genome 230 2.0 Array 1369291_at - -
          Rat Genome 230 2.0 Array 1384240_at - -
          Rat Genome 230 2.0 Array 1384240_PM_at - -
          Affymetrix HT Rat Focus Array 1369291_at - -
          Affymetrix HT Rat Focus Array 1384240_at - -
          Affymetrix HT Rat Focus Array 1384240_PM_at - -
          Affymetrix HT Rat Focus Array 1369291_PM_at - -
          Rat Expression Set 230 1384240_at - -
          Rat Expression Set 230 1384240_PM_at - -
          Rat Expression Set 230 1369291_at - -
          Rat Expression Set 230 1369291_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798194 - -
          Rat Genome U34 Set M74054_s_at - -
          Rat Genome U34 Set X62295cds_s_at - -
          Rat Genome U34 Set M86912exon_g_at - -
          Rat Genome U34 Set M86912exon_at - -
          Rat Genome U34 Set M74054_s_at - -
          Rat Genome U34 Set X62295cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21329950 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364972 - -
          RatRef-12 Expression BeadChip ILMN_56459 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline