Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02758803_s1
          See other GPR19 GE Assays ›
          Gene Symbol:
          Gpr19
          Gene Name:
          G protein-coupled receptor 19
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 168639930 - 168668333 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02758803_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080579.1 NP_542146.1 2 1361 95
          XM_006237520.2 XP_006237582.1 5 3544 95
          XM_006237521.2 XP_006237583.1 5 1825 95
          XM_006237522.2 XP_006237584.1 3 1370 95
          XM_006237523.2 XP_006237585.1 3 1617 95
          XM_006237524.1 XP_006237586.1 3 1527 95
          XM_008763379.1 XP_008761601.1 4 2277 95
          GenBank mRNA BC089971.1 - 2 1361 7315466 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr19

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135594 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005852 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90747 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866411 - -
          Affymetrix Rat Gene 1.0 ST Array 10866412 - -
          Affymetrix Rat Gene 1.1 ST Array 10866410 - -
          Rat Genome 230 2.0 Array 1382739_PM_at - -
          Rat Genome 230 2.0 Array 1382739_at - -
          Affymetrix HT Rat Focus Array 1382739_PM_at - -
          Affymetrix HT Rat Focus Array 1382739_at - -
          Rat Expression Set 230 1382739_at - -
          Rat Expression Set 230 1382739_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10866410 - -
          Rat Genome U34 Set U65417cds_at - -
          Rat Genome U34 Set rc_AI045718_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22112777 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354069 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline