Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02758918_s1
          See other RGD1560455 GE Assays ›
          Gene Symbol:
          RGD1560455
          Gene Name:
          similar to RIKEN cDNA A630033H20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 78258250 - 78284180 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02758918_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1560455
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106939.1 NP_001100409.1 2 1347 114
          XM_006257166.2 XP_006257228.1 2 1851 114
          XM_006257168.2 XP_006257230.1 2 1274 114
          XM_006257169.2 XP_006257231.1 2 1457 114

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064976 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161785 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177015 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064976 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064983 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064976 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064983 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064976 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064983 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P253253 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26621 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934638 - -
          Affymetrix Rat Gene 1.1 ST Array 10934637 - -
          Affymetrix Rat Clariom S Assay PSR0X00004612.rn.1 TC0X00000730.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10934637 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22150211 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline