Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02769414_s1
          See other MLLT11 GE Assays ›
          Gene Symbol:
          Mllt11
          Gene Name:
          myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 11
          Gene Aliases:
          RGD1305525
          Chromosome Location:
          Chr.2: 196392936 - 196402106 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02769414_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mllt11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013912.1 NP_001013934.1 2 167 65
          GenBank mRNA BC087583.1 - 2 167 7316660 65
          BC094215.1 - 2 167 7318415 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mllt11

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          positive regulation of apoptotic process
          positive regulation of transcription, DNA-templated
          positive regulation of mitochondrial depolarization
          positive regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway
          intrinsic apoptotic signaling pathway
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112980 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014717 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014717 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014717 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P203793 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46603 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72921 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7311 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54905 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34789 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8790 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824968 - -
          Affymetrix Rat Gene 1.0 ST Array 10824967 - -
          Affymetrix Rat Gene 1.0 ST Array 10824966 - -
          Affymetrix Rat Gene 1.1 ST Array 10824965 - -
          Rat Genome 230 2.0 Array 1371692_at - -
          Rat Genome 230 2.0 Array 1371692_PM_at - -
          Affymetrix HT Rat Focus Array 1371692_PM_at - -
          Affymetrix HT Rat Focus Array 1371692_at - -
          Affymetrix Rat Clariom S Assay PSR0200028710.rn.1 TC0200004202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028711.rn.1 TC0200004202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028705.rn.1 TC0200004202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028707.rn.1 TC0200004202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028706.rn.1 TC0200004202.rn.1 RTA TC
          Rat Expression Set 230 1371692_at - -
          Rat Expression Set 230 1371692_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824965 - -
          Rat Genome U34 Set rc_AI009661_at - -
          Rat Genome U34 Set rc_AI071949_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650165 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline