Search
Search
| Gene Symbol: Ccdc71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024904.1 | NP_001020075.1 | 2 | 1267 | 66 | |
| XM_006243832.1 | XP_006243894.1 | 2 | 1296 | 66 | ||
| XM_006243835.2 | XP_006243897.1 | 2 | 1336 | 66 | ||
| GenBank mRNA | BC098020.1 | - | 2 | 1267 | 7455232 | 66 |
Set Membership: |
|