Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02769537_s1
          See other MAGEB16 GE Assays ›
          Gene Symbol:
          Mageb16
          Gene Name:
          melanoma antigen family B, 16
          Gene Aliases:
          Snca
          Chromosome Location:
          Chr.X: 46977655 - 47026377 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02769537_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mageb16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025013.1 NP_001020184.1 3 1167 71
          XM_006256976.1 XP_006257038.1 4 1227 71
          XM_006256977.2 XP_006257039.1 5 1290 71
          GenBank mRNA BC079208.1 - 3 1167 7131795 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mageb16

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P215646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189986 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P215646 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P215646 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P215646 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P215646 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44964 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81849 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92092 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938199 - -
          Affymetrix Rat Gene 1.0 ST Array 10938200 - -
          Affymetrix Rat Gene 1.0 ST Array 10938198 - -
          Affymetrix Rat Gene 1.1 ST Array 10938197 - -
          Rat Genome 230 2.0 Array 1393645_PM_at - -
          Rat Genome 230 2.0 Array 1393645_at - -
          Affymetrix HT Rat Focus Array 1393645_at - -
          Affymetrix HT Rat Focus Array 1393645_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00011410.rn.1 TC0X00001763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00011406.rn.1 TC0X00001763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00011407.rn.1 TC0X00001763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00011408.rn.1 TC0X00001763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00011405.rn.1 TC0X00001763.rn.1 RTA TC
          Rat Expression Set 230 1393645_at - -
          Rat Expression Set 230 1393645_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938197 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline