Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02769730_g1
          See other RHOG GE Assays ›
          Gene Symbol:
          Rhog
          Gene Name:
          ras homolog family member G
          Gene Aliases:
          Arhg
          Chromosome Location:
          Chr.1: 167336147 - 167347708 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02769730_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhog
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037195.1 NP_001032272.1 2 371 68
          XM_006229882.2 XP_006229944.1 3 512 68
          XM_006229883.2 XP_006229945.1 2 618 68
          GenBank mRNA BC107943.1 - 2 371 7389075 68
          FQ212590.1 - 2 411 68

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rhog

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          nucleocytoplasmic transport
          small GTPase mediated signal transduction
          Rho protein signal transduction
          metabolic process
          Rac protein signal transduction
          actin cytoskeleton organization
          positive regulation of transcription, DNA-templated
          cell chemotaxis
          positive regulation of establishment of protein localization to plasma membrane
          regulation of ruffle assembly
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P046482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585343 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91690 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149655 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046482 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046482 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P046482 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P777045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724210 - -
          Affymetrix Rat Gene 1.0 ST Array 10724209 - -
          Affymetrix Rat Gene 1.1 ST Array 10724208 - -
          Rat Genome 230 2.0 Array 1389227_at - -
          Rat Genome 230 2.0 Array 1389227_PM_at - -
          Affymetrix HT Rat Focus Array 1389227_PM_at - -
          Affymetrix HT Rat Focus Array 1389227_at - -
          Affymetrix Rat Clariom S Assay PSR0100049224.rn.1 TC0100006179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049222.rn.1 TC0100006179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049228.rn.1 TC0100006179.rn.1 RTA TC
          Rat Expression Set 230 1389227_PM_at - -
          Rat Expression Set 230 1389227_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724208 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20989899 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364060 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline