Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02769958_s1
          See other GPRASP1 GE Assays ›
          Gene Symbol:
          Gprasp1
          Gene Name:
          G protein-coupled receptor associated sorting protein 1
          Gene Aliases:
          pips
          Chromosome Location:
          Chr.X: 106252753 - 106313854 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02769958_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gprasp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134386.1 NP_599213.1 2 5967 129
          XM_006257264.2 XP_006257326.1 9 4952 129
          XM_006257265.2 XP_006257327.1 9 5775 129
          XM_006257267.2 XP_006257329.1 8 5644 129
          XM_008773392.1 XP_008771614.1 11 5159 129
          XM_008773393.1 XP_008771615.1 10 5083 129
          XM_008773394.1 XP_008771616.1 11 5163 129
          XM_008773395.1 XP_008771617.1 11 5203 129
          XM_008773396.1 XP_008771618.1 9 5737 129
          XM_008773397.1 XP_008771619.1 9 5011 129
          XM_008773398.1 XP_008771620.1 9 5010 129
          GenBank mRNA AB051807.1 - 2 5967 129

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gprasp1

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          endosome to lysosome transport
          G-protein coupled receptor catabolic process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P459210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P459210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P459210 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P459210 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P459210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13138 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940431 - -
          Affymetrix Rat Gene 1.1 ST Array 10940429 - -
          Rat Genome 230 2.0 Array 1387148_at - -
          Rat Genome 230 2.0 Array 1387148_PM_at - -
          Affymetrix HT Rat Focus Array 1387148_PM_at - -
          Affymetrix HT Rat Focus Array 1387148_at - -
          Affymetrix Rat Clariom S Assay PSR0X00017206.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017224.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017205.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017207.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017210.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017211.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017225.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017212.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017208.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017204.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017226.rn.1 TC0X00002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00017223.rn.1 TC0X00002689.rn.1 RTA TC
          Rat Expression Set 230 1387148_PM_at - -
          Rat Expression Set 230 1387148_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940429 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20727637 - -
          ABI Rat Genome Survey Microarray 21338731 - -
          ABI Rat Genome Survey Microarray 22040933 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354368 - -
          RatRef-12 Expression BeadChip ILMN_63182 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline